Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170539.1 | NP_001164010.1 | 7 - 8 | 1179 | 71 | |
GenBank mRNA | BC166740.1 | - | 7 - 8 | 1179 | 7937421 | 71 |
FQ210494.1 | - | 7 - 8 | 1180 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |