Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170539.1 | NP_001164010.1 | 8 - 9 | 1226 | 98 | |
GenBank mRNA | BC166740.1 | - | 8 - 9 | 1226 | 7937421 | 98 |
FQ210494.1 | - | 8 - 9 | 1227 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |