Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110838.1 | NP_001104308.1 | 7 - 8 | 905 | 77 | |
XM_006244192.2 | XP_006244254.1 | 7 - 8 | 1017 | 77 | ||
XM_006244193.2 | XP_006244255.1 | 7 - 8 | 1126 | 77 | ||
XM_006244194.2 | XP_006244256.1 | 7 - 8 | 1042 | 77 | ||
XM_006244195.2 | XP_006244257.1 | 7 - 8 | 1010 | 77 | ||
XM_008766717.1 | XP_008764939.1 | 7 - 8 | 1042 | 77 | ||
XM_008766718.1 | XP_008764940.1 | 6 - 7 | 1151 | 77 | ||
XM_008766719.1 | XP_008764941.1 | 6 - 7 | 909 | 77 | ||
XM_008766720.1 | XP_008764942.1 | 6 - 7 | 874 | 77 | ||
XM_008766721.1 | XP_008764943.1 | 6 - 7 | 906 | 77 | ||
XM_008766722.1 | XP_008764944.1 | 7 - 8 | 1016 | 77 | ||
XM_008766723.1 | XP_008764945.1 | 6 - 7 | 1021 | 77 | ||
XM_008766724.1 | XP_008764946.1 | 7 - 8 | 934 | 77 | ||
XM_008766725.1 | XP_008764947.1 | 7 - 8 | 938 | 77 | ||
XM_008766726.1 | XP_008764948.1 | 6 - 7 | 802 | 77 | ||
GenBank mRNA | FQ210480.1 | - | 7 - 8 | 919 | 77 | |
FQ211645.1 | - | 7 - 8 | 990 | 77 | ||
FQ215070.1 | - | 7 - 8 | 919 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |