Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013954.1 | NP_001013976.1 | 4 - 5 | 1081 | 100 | |
XM_008766249.1 | XP_008764471.1 | 3 - 4 | 1025 | 100 | ||
XM_008766250.1 | XP_008764472.1 | 2 - 3 | 906 | 100 | ||
XM_008766251.1 | XP_008764473.1 | 4 - 5 | 1116 | 100 | ||
XM_008766252.1 | XP_008764474.1 | 3 - 4 | 969 | 100 | ||
XM_008766253.1 | XP_008764475.1 | 3 - 4 | 1412 | 100 | ||
XM_008766255.1 | XP_008764477.1 | 4 - 5 | 1141 | 100 | ||
XM_008766256.1 | XP_008764478.1 | 3 - 4 | 947 | 100 | ||
XM_008766257.1 | XP_008764479.1 | 3 - 4 | 1586 | 100 | ||
XM_008766258.1 | XP_008764480.1 | 3 - 4 | 4610 | 100 | ||
XM_008766259.1 | XP_008764481.1 | 3 - 4 | 1026 | 100 | ||
GenBank mRNA | BC083657.1 | - | 2 - 3 | 902 | 98 | |
BC085805.1 | - | 4 - 5 | 1081 | 7112021 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |