Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191883.1 | NP_001178812.1 | 1 - 2 | 156 | 63 | |
XM_006243802.2 | XP_006243864.1 | 2 - 3 | 652 | 63 | ||
XM_006243803.2 | XP_006243865.1 | 2 - 3 | 651 | 63 | ||
GenBank mRNA | AW533143.1 | - | 1 - 2 | 86 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |