Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007685.1 | NP_001007686.1 | 1 - 2 | 117 | 61 | |
GenBank mRNA | BC083777.1 | - | 1 - 2 | 117 | 7193594 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |