Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005383.1 | NP_001005383.1 | 10 - 11 | 855 | 74 | |
XM_006243715.2 | XP_006243777.1 | 11 - 12 | 1157 | 74 | ||
XM_006243716.2 | XP_006243778.1 | 11 - 12 | 1161 | 74 | ||
GenBank mRNA | AY580164.1 | - | 10 - 11 | 855 | 74 | |
BC078930.1 | - | 11 - 12 | 918 | 7107165 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |