Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106862.1 | NP_001100332.1 | 1 - 2 | 188 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |