Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 7 - 8 | 1057 | 81 | |
XM_006243872.2 | XP_006243934.1 | 7 - 8 | 1056 | 81 | ||
XM_006243880.2 | XP_006243942.1 | 3 - 4 | 668 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |