Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766605.1 | XP_008764827.1 | 13 - 14 | 5446 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |