Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001055346.5 | XP_001055346.3 | 2 - 3 | 135 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |