Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106860.1 | NP_001100330.1 | 3 - 4 | 251 | 64 | |
XM_008766512.1 | XP_008764734.1 | 4 - 5 | 416 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |