Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_236761.3 | XP_236761.1 | 2 - 3 | 965 | 93 |
Gene Symbol: RGD1559995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_235194.3 | XP_235194.1 | 1 | 345 | 93 |
Gene Symbol: RGD1564306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_229957.4 | XP_229957.2 | 1 | 367 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |