Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013252.1 | NP_001013270.1 | 2 - 3 | 272 | 113 | |
GenBank mRNA | AY325214.1 | - | 2420 | 113 | ||
AY325230.1 | - | 2423 | 113 | |||
AY325241.1 | - | 2420 | 113 | |||
BC098951.1 | - | 264 | 7324816 | 113 | ||
BF525252.1 | - | 278 | 113 | |||
BQ204036.1 | - | 5 - 6 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |