Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Birc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021752.2 | NP_068520.2 | 3 - 4 | 1683 | 79 | |
XM_006242494.2 | XP_006242556.1 | 4 - 5 | 2160 | 79 | ||
XM_006242495.2 | XP_006242557.1 | 4 - 5 | 2361 | 79 | ||
GenBank mRNA | AF183431.1 | - | 3 - 4 | 1041 | 79 | |
AF190020.1 | - | 3 - 4 | 2055 | 79 | ||
BC062055.1 | - | 3 - 4 | 1683 | 5598786 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |