Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080580.2 | NP_542147.1 | 1 - 2 | 104 | 138 | |
GenBank mRNA | BC081741.1 | - | 1 - 2 | 104 | 7110109 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |