Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008317.1 | NP_001008318.1 | 5 - 6 | 546 | 142 | |
GenBank mRNA | AY158662.1 | - | 5 - 6 | 546 | 142 | |
FQ211910.1 | - | 640 | 142 | |||
FQ216126.1 | - | 5 - 6 | 600 | 142 | ||
FQ227188.1 | - | 5 - 6 | 606 | 142 | ||
FQ234126.1 | - | 5 - 6 | 607 | 142 | ||
FQ234429.1 | - | 5 - 6 | 647 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |