Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012213.1 | NP_001012213.1 | 5 - 6 | 571 | 67 | |
XM_006253645.2 | XP_006253707.1 | 5 - 6 | 782 | 67 | ||
XM_008771502.1 | XP_008769724.1 | 5 - 6 | 543 | 67 | ||
GenBank mRNA | BC078935.1 | - | 5 - 6 | 571 | 7108166 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |