Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198050.1 | NP_932167.1 | 13 - 14 | 3074 | 78 | |
XM_006253556.2 | XP_006253618.1 | 13 - 14 | 3218 | 78 | ||
XM_006253557.2 | XP_006253619.1 | 12 - 13 | 3074 | 78 | ||
XM_008771424.1 | XP_008769646.1 | 13 - 14 | 3242 | 78 | ||
GenBank mRNA | BC079385.1 | - | 7 - 8 | 1066 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |