Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198050.1 | NP_932167.1 | 5 - 6 | 1742 | 107 | |
XM_006253556.2 | XP_006253618.1 | 5 - 6 | 1886 | 107 | ||
XM_008771424.1 | XP_008769646.1 | 5 - 6 | 1910 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |