Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057141.1 | NP_476482.1 | 9 - 10 | 976 | 80 | |
XM_006253552.2 | XP_006253614.1 | 11 - 12 | 1383 | 80 | ||
XM_006253553.2 | XP_006253615.1 | 10 - 11 | 1079 | 80 | ||
XM_006253554.2 | XP_006253616.1 | 11 - 12 | 1383 | 80 | ||
XM_006253555.2 | XP_006253617.1 | 10 - 11 | 1311 | 80 | ||
GenBank mRNA | BC061867.1 | - | 10 - 11 | 1063 | 5598872 | 80 |
D17711.1 | - | 9 - 10 | 976 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |