Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253552.2 | XP_006253614.1 | 2 - 3 | 400 | 121 | |
XM_006253554.2 | XP_006253616.1 | 2 - 3 | 400 | 121 | ||
XM_006253555.2 | XP_006253617.1 | 2 - 3 | 400 | 121 | ||
GenBank mRNA | BC061867.1 | - | 1 - 2 | 80 | 5598872 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |