Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191691.2 | NP_001178620.2 | 7 - 8 | 1944 | 84 | |
GenBank mRNA | AY157999.1 | - | 1 - 2 | 59 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |