Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037362.1 | NP_001032439.1 | 3 - 4 | 168 | 82 | |
XM_006253563.2 | XP_006253625.1 | 2 - 3 | 299 | 82 | ||
GenBank mRNA | BC107923.1 | - | 3 - 4 | 168 | 7444531 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |