Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Penk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237835.1 | XP_006237897.1 | 1 - 2 | 175 | 66 | |
GenBank mRNA | FQ213928.1 | - | 1 - 2 | 155 | 66 | |
M28263.1 | - | 1 - 2 | 43 | 66 | ||
S49491.1 | - | 1 - 2 | 113 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |