Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106572.1 | NP_001100042.1 | 5 - 6 | 1102 | 79 | |
XM_006234074.2 | XP_006234136.1 | 6 - 7 | 1310 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |