Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108953.1 | NP_001102423.1 | 1 - 2 | 78 | 117 | |
XM_006234101.2 | XP_006234163.1 | 1 - 2 | 174 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |