Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134580.1 | NP_001128052.1 | 7 - 8 | 634 | 150 | |
XM_008761762.1 | XP_008759984.1 | 4 - 5 | 2480 | 150 | ||
XM_008761763.1 | XP_008759985.1 | 5 - 6 | 3130 | 150 | ||
XM_008761764.1 | XP_008759986.1 | 4 - 5 | 2946 | 150 | ||
XM_008761765.1 | XP_008759987.1 | 6 - 7 | 3250 | 150 | ||
XM_008761766.1 | XP_008759988.1 | 3 - 4 | 2420 | 150 | ||
XM_008761767.1 | XP_008759989.1 | 3 - 4 | 2886 | 150 | ||
XM_008761768.1 | XP_008759990.1 | 3 - 4 | 1930 | 150 | ||
XM_008761769.1 | XP_008759991.1 | 4 - 5 | 2386 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |