Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008311.3 | NP_001008312.1 | 1 - 2 | 392 | 100 | |
GenBank mRNA | BC086596.1 | - | 1 - 2 | 392 | 7311836 | 100 |
DQ119821.1 | - | 320 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |