Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139090.1 | NP_620790.1 | 2 - 3 | 454 | 112 | |
GenBank mRNA | U35025.1 | - | 2 - 3 | 339 | 112 | |
U69702.1 | - | 2 - 3 | 454 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |