Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108581.1 | NP_001102051.1 | 3 - 4 | 539 | 75 | |
XM_006234155.2 | XP_006234217.1 | 3 - 4 | 513 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |