Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012086.1 | NP_001012086.1 | 4 - 5 | 432 | 63 | |
XM_006234202.2 | XP_006234264.1 | 5 - 6 | 406 | 63 | ||
GenBank mRNA | BC088207.1 | - | 4 - 5 | 432 | 7381695 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |