Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037649.1 | NP_001032738.1 | 2 - 3 | 157 | 95 | |
XM_006234167.2 | XP_006234229.1 | 2 - 3 | 221 | 95 | ||
XM_006234168.2 | XP_006234230.1 | 2 - 3 | 1019 | 95 | ||
XM_008761743.1 | XP_008759965.1 | 2 - 3 | 1023 | 95 | ||
XM_008761744.1 | XP_008759966.1 | 2 - 3 | 225 | 95 | ||
GenBank mRNA | BC107648.1 | - | 2 - 3 | 157 | 7930358 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |