Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdsub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014179.1 | NP_001014201.1 | 1 - 2 | 174 | 86 | |
GenBank mRNA | BC089856.1 | - | 1 - 2 | 174 | 7381399 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |