Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009684.1 | NP_001009684.1 | 2 - 3 | 383 | 83 | |
GenBank mRNA | BC088191.1 | - | 2 - 3 | 383 | 7372031 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |