Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250622.2 | XP_006250684.1 | 4 - 5 | 2559 | 84 | |
GenBank mRNA | L11354.1 | - | 1 - 2 | 120 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |