Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082540.1 | NP_001076009.1 | 2 - 3 | 562 | 139 | |
NM_024404.2 | NP_077380.2 | 2 - 3 | 562 | 139 | ||
GenBank mRNA | AB046615.1 | - | 2 - 3 | 326 | 139 | |
AB046617.1 | - | 2 - 3 | 277 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |