Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004275.1 | NP_001004275.1 | 7 - 8 | 919 | 126 | |
XM_006250667.2 | XP_006250729.1 | 6 - 7 | 829 | 126 | ||
XM_006250668.2 | XP_006250730.1 | 7 - 8 | 965 | 126 | ||
GenBank mRNA | BC079384.1 | - | 7 - 8 | 919 | 7111425 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |