Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034008.1 | NP_001029180.1 | 1 - 2 | 90 | 84 | |
GenBank mRNA | BC160817.1 | - | 1 - 2 | 69 | 7454028 | 84 |
CB758330.1 | - | 1 - 2 | 147 | 84 | ||
CV119475.1 | - | 1 - 2 | 68 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |