Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029920.1 | NP_001025091.1 | 3 - 4 | 548 | 100 | |
GenBank mRNA | BC098063.1 | - | 3 - 4 | 548 | 7458419 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |