Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031339.1 | NP_112629.1 | 3 - 4 | 1386 | 144 | |
XM_006252669.1 | XP_006252731.1 | 2 - 3 | 1056 | 144 | ||
GenBank mRNA | AB019366.1 | - | 3 - 4 | 1386 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |