Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107297.1 | NP_001100767.1 | 1 - 2 | 160 | 85 | |
XM_006252749.2 | XP_006252811.1 | 1 - 2 | 90 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |