Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138860.1 | NP_620215.1 | 4 - 5 | 823 | 70 | |
GenBank mRNA | AY072689.1 | - | 4 - 5 | 823 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |