Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009271.1 | NP_001009271.1 | 3 - 4 | 533 | 105 | |
XM_008770986.1 | XP_008769208.1 | 3 - 4 | 2575 | 105 | ||
GenBank mRNA | AY569011.1 | - | 3 - 4 | 526 | 105 | |
BC088431.1 | - | 3 - 4 | 533 | 7124640 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |