Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009271.1 | NP_001009271.1 | 12 - 13 | 1392 | 65 | |
XM_008770986.1 | XP_008769208.1 | 12 - 13 | 3434 | 65 | ||
GenBank mRNA | AY569011.1 | - | 12 - 13 | 1385 | 65 | |
BC088431.1 | - | 12 - 13 | 1392 | 7124640 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |