Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053791.2 | NP_446243.1 | 1 - 2 | 70 | 89 | |
GenBank mRNA | AF226608.1 | - | 1 - 2 | 68 | 89 | |
BC097296.1 | - | 1 - 2 | 70 | 7445776 | 89 | |
DQ329279.1 | - | 1 - 2 | 68 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |