Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253849.2 | XP_006253911.1 | 5 - 6 | 637 | 113 | |
GenBank mRNA | FQ210031.1 | - | 5 - 6 | 509 | 113 | |
FQ227425.1 | - | 5 - 6 | 505 | 113 | ||
FQ229485.1 | - | 5 - 6 | 519 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |