Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145786.1 | NP_665729.1 | 12 - 13 | 1322 | 130 | |
XM_006231016.2 | XP_006231078.1 | 11 - 12 | 1214 | 130 | ||
XM_008760216.1 | XP_008758438.1 | 12 - 13 | 1428 | 130 | ||
GenBank mRNA | AY032664.1 | - | 11 - 12 | 1186 | 130 | |
AY032665.1 | - | 12 - 13 | 1322 | 130 | ||
BC081718.1 | - | 12 - 13 | 1313 | 7112534 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |