Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Men1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019208.3 | NP_062081.1 | 3 - 4 | 1187 | 112 | |
XM_006230769.2 | XP_006230831.1 | 4 - 5 | 905 | 112 | ||
XM_006230770.2 | XP_006230832.1 | 4 - 5 | 996 | 112 | ||
XM_006230771.2 | XP_006230833.1 | 4 - 5 | 906 | 112 | ||
XM_006230772.2 | XP_006230834.1 | 4 - 5 | 964 | 112 | ||
GenBank mRNA | AB023400.1 | - | 3 - 4 | 1254 | 112 | |
AF130369.1 | - | 3 - 4 | 1162 | 112 | ||
AF130370.1 | - | 3 - 4 | 807 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |