Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106306.1 | NP_001099776.1 | 11 - 12 | 1927 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |